Hanging Cell Models
# de catalogue 470002-684
Fournisseur: Science First
Spécifications
- Description:Plant Cell
- No. of Keyed Items:23
- Cat. no.:470002-684
- Length:46 cm
- Width:61 cm
- Supplier No.:B 142
- No. of Parts:1
Spécifications
A propos de cet article
For Demonstration And Display
These high visibility large models are vacuum formed sheets of heavy plastic, molded to show in relief all the known organelles and inclusions of these two important cells. Twenty structures are included in the animal cell; twenty-three in the plant cell. All are identified by the accompanying key cards. Hand painted in brilliant color and are fitted with an easel back which permits the model to sit upright on a surface of hang on a wall with equal ease.
Customers Who Bought This Also Bought