Caustic Spill Clean Up Kit
# de catalogue 470127-674
Fournisseur: Ward's Science
Spécifications
- Description:Caustic Spill Clean Up Kit
- Cat. no.:470127-674
- Supplier No.:IS5002
Spécifications
A propos de cet article
Safe and Easy Clean-Up.
- Handy compact design
- Contains everything to safely contain the spill
Safely respond to accidental caustic spills in your chemistry lab with this complete, ready-to-use kit. Includes one bag of Vermiculite absorbent, one bag of neutralizing mixture, two 9" x 16" polybags, two blank shipping tags, two vinyl exam gloves, twist ties, and a dust pan and brush set. A detailed instruction sheet/SDS is provided.
Customers Who Bought This Also Bought