Spécifications
- Length:2"
- Width:3"
- Description:Cat Brain Anatomy
- Height:15.25"
- Cat. no.:470000-342
- Supplier No.:WARD470000-342
- No. of Parts:1
Spécifications
A propos de cet article
Museum mounts are the ideal supplement for classroom dissection.
- High quality display mount
- Dissection reference for students
- Quiz mechanism for teachers
Five different views and exceptional inner details.
Museum Mounts are expertly crafted by our own Laboratory Technicians to show external and internal anatomy in great detail. Specimens are encased in acrylic with formalin for indefinite preservation.
Museum mounts can be used as a reference for teachers and students or as a quiz mechanism by covering up the key on the back of the case.
Key includes: 1. Eyeball; 2. Cerebrum; 3. Cerebellum; 4. Olfactory Lobe; 5. Corpus Callosum; 6. Medulla Oblongata; 7. Pituitary Gland; 8. Pons; 9. Gray Matter; 10. Cortex; 11. Third Ventricle; 12. Thalamus.