Casting Kit
# de catalogue 470015-346
Fournisseur: FORENSICS SOURCE
Spécifications
- Description:Casting Kit
- No. of Lab Groups:5
- Cat. no.:470015-346
- Supplier No.:TRXT-KIT
Spécifications
A propos de cet article
Make Detailed Casts Using Professional-Grade Materials
- Same materials used by Forensic Experts
- Quick-drying material
The kit comes with five 1-gallon plastic resealable bags with 1 lb. of casting material in each, one measuring cup, one small bottle of hardening solution, a 12" ruler, and instructions.
Available to U.S. customers only, similar product will ship elsewhere.
Customers Who Bought This Also Bought