Cartilage Knife
# de catalogue 470006-524
Fournisseur: Dunrite Instruments
Spécifications
- Length:165 mm (6.5")
- Description:Cartilage Knife
- Material:Forged Steel
- Cat. no.:470006-524
- Supplier No.:140930
Spécifications
A propos de cet article
Designed for heavy use and rough cutting, the forged steel knife has a serrated handle to grip, and a large, 2" blade to cut through tough cartilage.
Caution: These instruments are intended for training and educational use only. They are not intended for human or animal procedures.
Customers Who Bought This Also Bought