Carteria
# de catalogue 470179-720
Fournisseur: Avantor
Spécifications
- Description:Carteria (Small)
- No. of Students:30
- Cat. no.:470179-720
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470179-720
Spécifications
A propos de cet article
Freshwater. Unicellular; large cells, with four flagella. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought