GPI Anatomicals® Canine Jaw Model
# de catalogue 470029-460
Fournisseur: GPI ANATOMICALS
Small Business Enterprise
Spécifications
- Description:Canine Jaw
- Height:5 cm
- No. of Keyed Items:16
- Cat. no.:470029-460
- Length:12 cm
- Width:7 cm
- Supplier No.:9195
- No. of Parts:2
Spécifications
A propos de cet article
Realistic Model Of Canine Dental Health
Open, close, and separate this average size canine jaw for up-close examination of normal dog teeth and common dental pathologies. While healthy teeth are depicted on the right side of the jaw, diseased and damaged teeth appear on the left, illustrating fractured canine, periodontal disease, tarter accumulation, plaque, gingivitis, worn incisors, retained deciduous tooth, gingival recession and missing premolar.
Customers Who Bought This Also Bought