GPI Anatomicals® Canine 4-Stage Knee Model
Fournisseur: GPI ANATOMICALS
Certificats
A propos de cet article
Demonstrate Osteoarthritis
Illustrate degenerative joint disease with this life-size set of four models. Models depict a disease-free (normal) knee highlighting important structures including ligaments, tendons and patella. The progressive degeneration of the articular surfaces and the development of osteophytes or bone spurs are demonstrated in the other three models.
Customers Who Bought This Also Bought