Candle Holder
# de catalogue 470006-942
Fournisseur: ELECTRO MECHANICAL INSTRUMENTS
Spécifications
- Description:Candle Holder
- Cat. no.:470006-942
- Supplier No.:6015
Spécifications
A propos de cet article
Perfect for Your Meter Stick Optics Bench.
This special bracket holds a single candle on a meter stick for use as a light source. Paraffin candles are available separately.
Customers Who Bought This Also Bought