Butterfy/Insect Net with Bamboo Handle
# de catalogue 470103-676
Fournisseur: ROLF C. HAGEN (USA) CORP.
Spécifications
- Length:5"
- Width:4"
- Description:Dip Net
- Cat. no.:470103-676
- Supplier No.:11264
Spécifications
A propos de cet article
Inexpensive insect collecting net with bamboo handle.
- Economical
- Bamboo handle
Handle measures 47.25".
Customers Who Bought This Also Bought