Bullfrog Skull
# de catalogue 470027-944
Fournisseur: Ward's Science
Spécifications
- Description:Bullfrog Skull
- Height:1"
- Cat. no.:470027-944
- Length:2"
- Width:2.25"
- Supplier No.:WARD470027-944
- No. of Parts:1