ContactUsLinkComponent
Brownlee Electrolysis Appraratus
Brownlee Electrolysis Appraratus
# de catalogue 470017-826
Fournisseur:  VWR International
BROWNLEE ELECTROLYSIS APPARATUS
Brownlee Electrolysis Appraratus
# de catalogue 470017-826
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Brownlee Electrolysis Apparatus
  • Cat. no.:
    470017-826
  • Supplier No.:
    PH0941B
Spécifications
A propos de cet article
A Must Have For Any Chemistry Classroom or Lab.
  • Clear visual of electrolysis for every classroom
  • Can be used year after year
Increased functionality equips this device for accurate and precise use in chemistry experiments. The apparatus features front spring clips, which allow you to easily remove the test tubes while they remain submerged. For greater safety, the binding posts have also been re-positioned. This apparatus comes with both ungraduated and graduated test tubes and instructions. Size 14 cm x 15 cm x 5 cm. A simple beaker can be used as the battery jar for this apparatus, and is available separately.
Customers Who Bought This Also Bought