Brine Shrimp Eggs
Supplier: San Francisco Bay Brand
Certificates
About this item
Artemia sp.
- Eggs hatch in two days
- Economical
- Instructions included
An inexpensive way to produce plenty of brine shrimp for study or food, eggs hatch within two days of being placed in a salt solution. Culture instructions are included.
Customers Who Bought This Also Bought