Breadboard Jumpers
# de catalogue 470105-634
Fournisseur: GLOBAL SPECIALTIES SE
Spécifications
- Description:Jumpers, Breadboard, Assorted Lenghts50
- Cat. no.:470105-634
- Supplier No.:111-0044
Spécifications
A propos de cet article
Use this colorful, 350 piece assortment of 22 gauge solid wire jumpers to make connections on your breadboards. The cut and formed jumpers range from 0.1 to 5.0" in 14 different lengths in a storage case.
The flexible jumpers are ideal for breadboard use. The wire is flexible, but the ends are solid, making breadboarding quick and easy.
Customers Who Bought This Also Bought