Bravais Space Lattice Model Set
# de catalogue 470029-192
Fournisseur: Klinger
Spécifications
- Description:Bravais space lattice model set, 14 pieces
- Cat. no.:470029-192
- Supplier No.:KLINGER EDUCATION PROD CORP SE
Spécifications
A propos de cet article
Study the Atomic Arrangements That Make up Most Minerals
- 14 Models
- Sturdy Wooden Models
This large, sturdy set of 14 lattice-type models is crafted of wood spheres connected by metal rods, so it won’t break easily, even with frequent handling. Six vibrant colors identify each model with one of the six fundamental crystal systems.
Models:
Triclinic; monoclinic/base-centered; orthorhombic/base, face, body-centered; tetragonal/body-centered; rhombohedral; and hexagonal; cubic/face, body-centered.
Each model is clearly labeled. Size: up to 6".
Customers Who Bought This Also Bought