Brain, Composite, Mammal Slide
# de catalogue 470182-728
Fournisseur: Avantor
Spécifications
- Description:Brain, Composite, Mammal Slide
- Cat. no.:470182-728
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-728
- Orientation:Section
Spécifications
A propos de cet article
Compare and contrast brain function with this composite slide
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal cerebrum, cerebellum, and medulla.
Customers Who Bought This Also Bought