Bowen's Reaction Series Mineral Collection
Catalog # 470025-186
Supplier: Avantor
Specifications
- Description:Bowen’s reaction series mineral collection
- Cat. no.:470025-186
- Supplier No.:WARD470025-186
- No. of Samples:16
Specifications
About this item
Specimens illustrate important stages of mineral formation.
- Demonstrates the formation of minerals
- Highest quality minerals
- Hand picked by WARD's Geologists
Samples included in the collection are representative of Bowen’s Reaction Series, from both the continuous and discontinuous branches of minerals formed from cooling magma
The set is housed in a compartmented collection box
Contents:
A chart detailing the reaction series, 8¹/₂×11" in size
Identification list
16 numbered samples, 1×1", albite, andesine, anorthite, augite, biotite, bytownite, enstatite, hornblende, labradorite, muscovite, oligoclase, olivine, orthoclase (2), phlogopite, quartz
Ordering information: Due to mineral availability, some minerals may be substituted.
Customers Who Bought This Also Bought