Glass Bottles with Screw Cap
Fournisseur: J.G. FINNERAN ASSOCIATES, INC. SE
Certificats
A propos de cet article
Bottles come with a plastic screw cap that has a protective inner lining. Use amber bottles for light-sensitive liquids.
Customers Who Bought This Also Bought