Borrelia burgdorferi Slide
# de catalogue 470177-206
Fournisseur: Avantor
Spécifications
- Description:Borrelia burgdorferi
- Cat. no.:470177-206
- Stain:Giemsa
- Supplier No.:WARD470177-206
- Orientation:Smear
Spécifications
A propos de cet article
Agent of Lyme Disease in Humans
- Identifying features clearly distinguishable
- Rigorous quality control standards
Helical coils. From culture.
Customers Who Bought This Also Bought