Borosilicate Glass Geissler Burets
Fournisseur: GSC International
Certificats
A propos de cet article
Student grade buret with fitted glass stopcock.
- Glass tubing is annealed to relieve stress
- Divisions are clearly marked
Burettes are made from the highest quality accurate bore tubing to provide a high degree of accuracy, reliability, and durability. Amber graduations. Includes a straight bore glass stopcock. Accuracy as per Class B.
Customers Who Bought This Also Bought