Human Bone, Ground Preparation Slide
# de catalogue 470177-628
Fournisseur: Avantor
Spécifications
- Description:Bone (Human), Ground Preparation
- Cat. no.:470177-628
- Stain:unstained
- Supplier No.:WARD470177-628
- Orientation:Cross Section
Spécifications
A propos de cet article
cross section, unstained, thin section
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Thin section showing Haversian canals, lacunae, and canaliculi.
Customers Who Bought This Also Bought