Boiling Stones, Glass Beads and Tamer Tabs™
Catalog # 470205-478
Supplier: Walter Stern
Specifications
- Description:Glass boiling beads
- Cat. no.:470205-478
- Packaging:450 g in plastic bottle
- Supplier No.:100G
Specifications
About this item
Solid glass beads, 6 mm.
Customers Who Bought This Also Bought