ContactUsLinkComponent
Blood Spatter Detectives Kit
Blood Spatter Detectives Kit
# de catalogue 470121-000
Fournisseur:  Kemtec
BLOOD SPATTER DETECTIVES KIT
Blood Spatter Detectives Kit
# de catalogue 470121-000
Fournisseur:  Kemtec
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Blood Spatter Detectives Lab Activity
  • No. of Lab Groups:
    12
  • Cat. no.:
    470121-000
  • Supplier No.:
    10-550
Spécifications
A propos de cet article
Comprehensive kit for education.
  • Grades 9-12
  • Connections to physics and chemistry
Complete over six different experiments in this in-depth activity kit. It includes use of a build it yourself spatter chamber for worry free, aggressive casting techniques. This process also teaches students the 16 general rules of blood spatter interpretation and investigative technology terms. Experiments include Distance Effect on Blood Spatter, Identifying Distance and angle of travel with Blood Spatter, and Investigating Blood Spatter with Luminol. A focus on student test knowledge and cross-curricular use of science and writing skills are also applied to creating a crime scene scenario. Includes materials for 12 setups.
Customers Who Bought This Also Bought