Ward's® Blood Components Model Set
# de catalogue 470029-436
Fournisseur: Wards
Spécifications
- Description:Blood Components Set
- No. of Keyed Items:8
- Cat. no.:470029-436
- Supplier No.:810601
- No. of Parts:8
Spécifications
A propos de cet article
Exceptional realism recreates details normally visible only under a microscope.
- Full Set of Blood Components on Individual Bases
- Detailed Guide Describes Each Cell Model and Its Function in the Circulatory System
- Designed By Wards
This set provides students with a detailed view of blood elements without a microscope. Cast in durable, lightweight plastic and hand painted under the direction of our staff anatomist for realistic features, the eight individual models consist of the erythrocyte, neutrophil, eosinophil, basophil, small lymphocyte, large lymphocyte, monocyte, and platelets.
The models are enlarged 10,000x and are rendered in a centimeter scale that equals micron dimensions of cells in blood smears.
Ordering information: The eight models are mounted on individual stands, and the set includes a teacher’s manual describing each model.
Customers Who Bought This Also Bought