Prepared Blood Agar Media
Fournisseur: Culture Media & Supplies
Certificats
A propos de cet article
Tryptic Soy Agar base with 5% Sheep Blood
- Excellent for cultivation of fastidious organisms
- Plate your organism onto Blood agar plates to test for hemolytic reaction
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.