Spécifications
- Description:Bitumen from Oil Sands Lab Activity
- No. of Lab Groups:10
- Cat. no.:470175-692
- Supplier No.:WARD470175-692
Spécifications
A propos de cet article
Introduction to Bitumen Extraction for Conversion to Crude Oil
- Demonstrates methods required to recover bitumen that is eventually produced as synthetic crude oil
- Initiates questions about fossil fuel energy use and environmental, social, and economical impacts of mining
- Materials included for ten lab setups
Using actual oil sands from the Athabasca oil sand deposits in Alberta, students will examine the nature and characteristics of oil sand and then formulate and test methods of bitumen extraction.