Biology Is Outdoors! A Comprehensive Resource For Studying School Environments
# de catalogue 470002-380
Fournisseur: Walch Publishing
Spécifications
- Description:Biology Is Outdoors!
- Cat. no.:470002-380
- No. of Pages:142
- Length:21 cm
- Width:27 cm
- Supplier No.:017976
Spécifications
A propos de cet article
Judith M. Hancock
Take your class on a field trip... to the schoolyard. Adaptable for any climate and geographic location, the activities on the reproducible pages of this book guide students as they study their immediate natural environment. Sections for teachers include discussion questions.
Customers Who Bought This Also Bought