Eisco Bicycle Wheel Gyroscope
Catalog # 470005-548
Supplier: VWR International
Specifications
- Description:Bicycle wheel gyroscope
- Cat. no.:470005-548
- Weight:3 kg (6.8 lbs.)
- Supplier No.:BI-SCP
- Diameter:50 cm (20")
Specifications
About this item
Easy to use because of its smaller diameter and simple means of hanging and loading, this gyroscope brings precession diagrams to life.
- 20" dia. rubber bicycle wheel with handles
- Demonstrates the conservation of angular momentum
- Can be suspended from a string (included)
Ideal for demonstrations in physical science and physics classrooms or laboratories. Inflate the bicycle tire for optimal results.
Customers Who Bought This Also Bought