Beyond Bead Biology: Molecular Biology Kit
Catalog # 470004-036
Supplier: Avantor
Specifications
- Description:Beyond Bead Biology: Molecular Biology Kit
- Cat. no.:470004-036
- Supplier No.:WARD470004-036
Specifications
About this item
Students use color-coded pop beads to visualize the structure of amino acid chains with this kit.
- Material for 10 Student Groups
- Teacher’s Guide with Lab Activities and Student Questions Included
- Topics Include Coenzymes, Substrates, Inhibitors, and More
- Tactile, Colorful Activities
- For Grades 10-12
Students work together to create the matching polymer of the DNA code they received by arranging beads in the proper sequence. Developed by high school biology teacher Mark Krotec, this comprehensive series of lab activities introduces topics such as coenzymes, substrates, inhibitors, mutations, DNA replication, recombinant DNA, PCR technique, mitosis, meiosis, and gene therapy.
This kit is for grades 10 through 12.
Customers Who Bought This Also Bought