Belenitella americana
# de catalogue 470225-108
Fournisseur: Avantor
Spécifications
- Description:Belemnitella americana
- Specimen Dimensions:2"
- Supplier No.:WARD470225-108
Spécifications
A propos de cet article
- Cephelopoda
- Classic Belemnite
- Cretaceous
- Average size 2"
Customers Who Bought This Also Bought