Ward's® Beating Sheet
# de catalogue 470018-452
Fournisseur: Avantor
Spécifications
- Description:Beating Sheet
- Cat. no.:470018-452
- Supplier No.:WARD470018-452
Spécifications
A propos de cet article
Collect Insects from Shrubbery
Don’t beat the bushes without our muslin beating sheet to collect the insects that live there. Insects can be easily seen and collected on this 36" square. The sheet is held open by diagonal support rods and is easily collapsible for carrying.
Customers Who Bought This Also Bought