Battery Operated Light Box
# de catalogue 470128-434
Fournisseur: VWR International
Spécifications
- Description:Battery Operated Light Box
- L×W×H:14 x 7 x 8 cm
- Weight:0.22 kg (0.5 lbs.)
- Supplier No.:PH0600-LED
Spécifications
A propos de cet article
This power–saving alternative to a traditional ray box is portable and durable.
The low power LED mounted inside this metal die cast raybox prevents heat buildup for added safety over extended periods of time. Unit operates on two AA batteries and is fitted with two 4 nm. sockets for the battery pack, which is easily mountable on the raybox. Includes collinating lens and slits with accomodating slots in integral siderails. Batteries not included.
- Led Prevents Heat Buildup
- Battery-Operated for Portability
- Sturdy Metal Housing
The low-power LED mounted inside this metal ray box prevents heat buildup for added safety over extended periods.
The unit operates on two AA batteries and is fitted with two 4 mm sockets for the battery pack, which is easily mountable on the ray box.
Ordering information: This box includes a collimating lens and slits with accommodating slots in integral side rails. Two AA batteries are required but not included.
Customers Who Bought This Also Bought