Basic Molecular Models Student Activity
# de catalogue 470122-968
Fournisseur: Lab-Aids
Spécifications
- Description:Basic Molecular Models Student Activity
- No. of Lab Groups:12
- Cat. no.:470122-968
- Includes:Teacher's Guide, Student Guides and Worksheets
- Supplier No.:130
Spécifications
A propos de cet article
Visualize and Understand How Elements Combine to Form Compounds
- Easy to assemble
- Demonstrate chemical bonding
- Requires one 50 minutes classroom period to complete
This introductory kit brings the basics into focus. As students are able to visualize “invisible” molecules, it becomes clear which have particular shapes and special properties and chemical behavior becomes more comprehensible.
Ordering information: Materials include 12 Packages each containing: 8 Hydrogen single electron atoms (white), 6 Chlorine tetrahedral centers (green), 4 Oxygen tetrahedral centers (blue), 2 Carbon tetrahedral centers (black), 2 Nitrogen tetrahedral centers (red), 1 Sulfur tetrahedral centers (yellow), 16 Covalent electron bonds, 1 Electron bond.
Customers Who Bought This Also Bought