Precision Measuring Set
# de catalogue 470144-808
Fournisseur: United Scientific Supplies
Spécifications
- Description:Precision Measuring Set
- Cat. no.:470144-808
- Supplier No.:PMSET04
Spécifications
A propos de cet article
Four High-Quality Instruments Housed in a Sturdy Plastic Storage Case.
- Complete kit is supplied in a fitted plastic case for easy stacking
- Transparent top allows for easy inspection
- Instructions included
Teach precision linear measurement, significant figures, and approximation with this unique kit. Give your students concrete experience using industrial tools and various unknowns. The kit includes a detailed instruction manual, a micrometer, vernier caliper, a knife-edge square, and a ruler. Unknowns to be measured include a 3/4" steel sphere, a round aluminum rod, a plugged aluminum tube, and a rectangular aluminum block.
Kit Contains:
1 Micrometer Caliper, 0-25mm x 0.01mm
1 Knife-Edge Square, 90°, 100mm x 70m
1 Vernier Caliper, 150mm x 0.02mm
1 Ruler, Steel, 15cm x 0.5mm
Customers Who Bought This Also Bought