Basic Hand-Held Stream Flowmeter
# de catalogue 470004-378
Fournisseur: GEOPACKS
Spécifications
- Description:Basic Hand-Held Stream Flowmeter
- Cat. no.:470004-378
- Supplier No.:ZMFP51
Spécifications
A propos de cet article
Lets You Control Location and Depth of Measurements
Using the three included elevation rods, you can measure velocity at fixed heights from 4–19" above the stream bed. The digital LCD meter is stored in a protective carrying pouch, and comes with a sturdy impeller stick that disassembles into four sections for easy storage, an instruction manual with laminated velocity calibration sheets and data sheets, and a compact weather-resistant case that conveniently holds the flowmeter, all additional parts, and two AAA batteries. Size of carrying case: 8" x 14" x 7". Total pole length: 3 1/2', extendable to 5' with an overall 7' lead. Weight: 2 lbs.
Customers Who Bought This Also Bought