Barr Bodies Present Slide
# de catalogue 470183-140
Fournisseur: Avantor
Spécifications
- Description:Barr Bodies Present
- Cat. no.:470183-140
- Stain:Biebrich and fast green
- Supplier No.:WARD470183-140
- Orientation:Smear
Spécifications
A propos de cet article
Illustrates Genetic "Femaleness" in Humans
- Identifying features clearly distinguishable
- Rigorous quality control standards
Smear of isolated squamous epithelial cells from the buccal cavity of a normal human female. Many nuclei show Barr bodies or drumstick chromosomes.
Customers Who Bought This Also Bought