Bacteriophage Model Set
# de catalogue 470005-104
Fournisseur: Science First
Spécifications
- Description:Bacteriophage Set
- No. of Keyed Items:8
- Cat. no.:470005-104
- Supplier No.:B-115
- No. of Parts:8
Spécifications
A propos de cet article
Individual Stages Of A Virus Destroying A Cell
To easily show how viruses attack the body to create infections, use the exceptional set of eight models depicting a T2 phage virus destroying a bacterium. Students can pick up and examine the individual, translucent models, enlarged 100,000X, to see what happens inside the bacterium at each stage. Each model represents a different step in the process:
Customers Who Bought This Also Bought