A propos de cet article
Two o strains of Escherichia coli; one exhibiting Ampicillin resistance, and one sensitive, but competent and ready to receive the plasmid that confers resistance.
- Incubation conditions 37 °C
Strain JM101: Includes the plasmid pUC8, which makes it resistant to Ampicillin.
Strain RRI: Can be transformed with pUC19 (available separately) plasmid to become resistant to Ampicillin and to metabolize lactose.
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.