Azurite Mineral Display
# de catalogue 470154-844
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Azurite Mineral Display, 2" x 3" to 3" x 4".
- Cat. no.:470154-844
- Supplier No.:3490014
Spécifications
A propos de cet article
China
Deep blue crystals, some as drusy coating on matrix or in small vugs; ave. size 2" x 3" to 3" x 4".
Customers Who Bought This Also Bought