ContactUsLinkComponent
Atmospheric Pressure Cups
Atmospheric Pressure Cups
# de catalogue 470110-932
Fournisseur:  VWR International
SUCTION CUPS SET2 PRESSURE DEMO
Atmospheric Pressure Cups
# de catalogue 470110-932
Fournisseur:  VWR International
Spécifications
  • Description:
    Atmospheric Pressure Cups
  • Cat. no.:
    470110-932
  • Supplier No.:
    PH0171
Spécifications
A propos de cet article
Demonstrate the Force of Air Pressure Without a Vacuum Pump
In this modern variant of a famous 17th-century experiment, students place the edges of two hemisphere cups together and squeeze them, forcing out the air between them. In a tug of war, students try to pull the cups apart-a surprisingly difficult task. Each of the 7.5cm-diameter rubber hemispheres has a two-finger tug handle and two release posts.
Customers Who Bought This Also Bought