A propos de cet article
Lightweight Chemical Splash Goggles are a step up from other bulky chemical splash goggles.
- Lightweight, 1⅘ oz.
- Lexa lenses
- Adjustable head strap
- Anti-fog
- Anti-scratch
These comfortable lightweight safety goggles feature Lexa lenses and an adjustable head strap.
The Lexa lenses are anti-fog, anti-scratch, anti-static, and absorb 99.9% UV protection.
470144-950 is latex-free goggles, including the straps (contains neoprene and polyester).