ContactUsLinkComponent
Ward's® Antibiotic Resistance Kit
Ward's® Antibiotic Resistance Kit
# de catalogue 470179-628
Fournisseur:  Avantor
WARDS ANTIBIOTIC RESISTANCE SET
Ward's® Antibiotic Resistance Kit
# de catalogue 470179-628
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    WARD’S Antibiotic Resistance Kit
  • Cat. no.:
    470179-628
  • Supplier No.:
    WARD470179-628
Spécifications
A propos de cet article
Using penicillin-impregnated discs on two plates, students grow one lawn each of two common, naturally occurring pathogen bacteria.
  • Students Compare Bacteria with and without Natural Resistance
  • Two Common Bacteria Included
  • Material for Five Lab Groups Included
  • Instructions Included
  • Penicillin-Impregnated Discs on Each Plate
The bacteria with natural resistance covers the entire plate with growth.
The bacteria without resistance shows a zone of inhibition (a ring of no growth around the disc).
Canada: Containment Level 2 – PHAC License required.
Ordering information: It includes Escherichia coli and Staphylococcus epidermidis tube cultures, material for five lab groups, and instructions.
Delivery: This kit includes perishable materials. Please specify delivery date.
Caution: Note: Living cultures are provided to reduce the laboratory preparation time. They can be stored in the refrigerator for up to 3 months before use.
Customers Who Bought This Also Bought