Annunciator Wire (Bell Wire)
# de catalogue 470005-926
Fournisseur: Arcor Electronics
Spécifications
- Description:Wire, Annunciator, 18 Gauge, 12 m 113 grams
- Cat. no.:470005-926
- Supplier No.:18A-SCIEN
Spécifications
A propos de cet article
A high quality PVC coated copper wire. AWG 18.
Customers Who Bought This Also Bought