Eisco® Animal Meiosis Model
# de catalogue 470111-022
Fournisseur: VWR International
Spécifications
- Description:Animal Meiosis
- Height:5 cm
- Cat. no.:470111-022
- Length:51 cm
- Width:46 cm
- Supplier No.:ZM0010
- No. of Parts:1
Spécifications
A propos de cet article
This set shows meiotic cellular division in every stage.
- Twelve Models per Set
- Models Mounted common base
- Models Shows Structures as Under Microscopes
Models are mounted on a common base.
The level of detail highlights all structures as they would appear under a microscope.
Customers Who Bought This Also Bought