ContactUsLinkComponent
DNA Fingerprinting After PCR Amplification
DNA Fingerprinting After PCR Amplification
# de catalogue 470126-638
Fournisseur:  Edvotek
AMPLIFICATION OF DNA FOR FINGERPRINTING
DNA Fingerprinting After PCR Amplification
# de catalogue 470126-638
Fournisseur:  Edvotek
Spécifications
  • Description:
    DNA Fingerprinting Lab Activity
  • No. of Lab Groups:
    8
  • Cat. no.:
    470126-638
  • Supplier No.:
    130
Spécifications
A propos de cet article
Apply Biotechnology to Forensics Investigations
  • Convenient Ready-To-Load samples
  • Inquiry-based experiment
  • No PCR reagents are included or required
  • Time required: ~45 minutes
Forensic DNA fingerprinting is quickly becoming a widely-accepted crime fighting tool, practiced by today's criminal investigation professionals. This inquiry-based experiment, based on a crime scene scenario, includes all the supplies necessary to analyze DNA extracted and already PCR amplified from fingerprints using gel electrophoresis. No PCR reagents are included or required. Ready-to-Load™ DNA is included for quick and easy sample loading.
Kit includes: instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Blue and FlashBlue™ stain, calibrated pipette, and microtipped transfer pipettes.
Required, but not included: electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, visualization system.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Customers Who Bought This Also Bought