Amber Latex Tubing
Fournisseur: Primeline
Certificats
A propos de cet article
Medium wall sulfur-free tubing.
- Smooth finish
- Translucent
This tubing is recommended for gas analysis or other applications where contamination must be avoided. Its clinging characteristics make it excellent for glass connections.
Customers Who Bought This Also Bought