ContactUsLinkComponent
Aluminum Overflow Can and Catch Bucket
Aluminum Overflow Can and Catch Bucket
# de catalogue 470201-426
Fournisseur:  VWR International
BUCKET ALUMINUM CATCH W/ HANDLE
Aluminum Overflow Can and Catch Bucket
# de catalogue 470201-426
Fournisseur:  VWR International
Spécifications
  • Description:
    Aluminum Catch Bucket
  • Cat. no.:
    470201-426
  • Supplier No.:
    PH0119D
Spécifications
A propos de cet article
This sturdy, seamless aluminum can measures 10 cm Ø.
A sturdy, seamless aluminum can measuring 7 cm in diameter. The top edge is flared and the spout is angled slightly. The aluminum catch bucket with handle measures 8 cm high and 5 cm in diameter.
  • Flared top edge
  • Angled spout
  • Sturdy
  • Seamless
The top edge is flared, and the spout is slightly angled.
The aluminum catch bucket with a handle measures 10 cm high and 10 cm Ø.
Customers Who Bought This Also Bought