Aluminum Coated Pith-Like Balls
# de catalogue 470104-434
Fournisseur: Sempco
Spécifications
- Description:Aluminum Coated Pith-Like Balls
- Cat. no.:470104-434
- Supplier No.:BH-9107
Spécifications
A propos de cet article
A thin aluminum coating produces great sensitivity to electrostatic charges. Styrofoam pith-like balls measure 8 mm in diameter and each has a silk suspension cord.
Packaging: Pack of six.
Customers Who Bought This Also Bought