Alnico Cylindrical Magnet
# de catalogue 470149-700
Fournisseur: United Scientific Supplies
Spécifications
- Description:Magnet, dia. 12.7 mm (0.5")
- Cat. no.:470149-700
- Length:75 mm (3")
- Supplier No.:COW-CP5
Spécifications
A propos de cet article
Strong alnico magnet with a magnetic pole on each end.
- Extremely strong magnet
- Approximate weight is 100 g
This versatile magnet, often referred to as a Cow Magnet, can be used for magnetic field demonstrations and a variety of other lab experiments.
Customers Who Bought This Also Bought