Alnico Bar Magnets
# de catalogue 470145-594
Fournisseur: Skywill Far East Industries
Spécifications
- Description:Alnico Bar magnets
- Cat. no.:470145-594
- Length:7.6
- Width:2 cm
- Supplier No.:2786000
Spécifications
A propos de cet article
Make magnetism fun and hands on with the alnico bar magnet.
Each is 3" in length and has marked North-South poles. Alnico material. Package of two.
Customers Who Bought This Also Bought